ContactUsLinkComponent
Bovine IGFBP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine IGFBP4 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4131-48
Supplier:  Antibodies.com
New Product
Bovine IGFBP4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of bovine IGFBP4 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4131-48
Supplier:  Antibodies.com