ContactUsLinkComponent
Mouse PSPN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse PSPN in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5443-48
Supplier:  Antibodies.com
New Product
Mouse PSPN ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f mouse PSPN in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5443-48
Supplier:  Antibodies.com