ContactUsLinkComponent
Bovine TRAP/CD40L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of bovine TRAP/CD40L in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4008-48
Supplier:  Antibodies.com
New Product
Bovine TRAP/CD40L ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of bovine TRAP/CD40L in serum, plasma, tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4008-48
Supplier:  Antibodies.com