ContactUsLinkComponent
Rat SOX2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SOX2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA4678-48
New Product
Rat SOX2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat SOX2 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA4678-48