ContactUsLinkComponent
Mouse Factor XII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Factor XII in plasma or other biological fluids. 1 * 48 Tests
  ANTIA5077-48
New Product
Mouse Factor XII ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of mouse Factor XII in plasma or other biological fluids. 1 * 48 Tests
  ANTIA5077-48