ContactUsLinkComponent
Lipopolysaccharides ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative det ermination of Lipopolysaccharides in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or othe 1 * 48 Tests
Catalog # ANTIA4716-48
Supplier:  ANTIBODIES.COM
New Product
Lipopolysaccharides ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative det ermination of Lipopolysaccharides in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or othe 1 * 48 Tests
Catalog # ANTIA4716-48
Supplier:  ANTIBODIES.COM