ContactUsLinkComponent
Human Tankyrase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Tankyrase in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA4861-96
New Product
Human Tankyrase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Tankyrase in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
  ANTIA4861-96