ContactUsLinkComponent
Rat Transferrin Receptor 2/TFR2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of rat Transferrin Receptor 2/TFR2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4320-48
Supplier:  Antibodies.com
New Product
Rat Transferrin Receptor 2/TFR2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of rat Transferrin Receptor 2/TFR2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4320-48
Supplier:  Antibodies.com