ContactUsLinkComponent
Rat Ki67 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ki67 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3837-48
Supplier:  Antibodies.com
New Product
Rat Ki67 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ki67 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3837-48
Supplier:  Antibodies.com