ContactUsLinkComponent
Human CD58 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD58 in serum, plasma, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3620-48
Supplier:  Antibodies.com
New Product
Human CD58 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human CD58 in serum, plasma, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3620-48
Supplier:  Antibodies.com