Rat Smad3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Smad3 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5639-48
Supplier: Antibodies.com