ContactUsLinkComponent
Mouse Enkephalin/ENK ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of mouse Enkephalin/ENK in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA3339-48
New Product
Mouse Enkephalin/ENK ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of mouse Enkephalin/ENK in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA3339-48