ContactUsLinkComponent
Human FADS3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FADS3 in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5391-48
New Product
Human FADS3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FADS3 in tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5391-48