ContactUsLinkComponent
Porcine Prolactin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of porcine Prolactin in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA3884-48
New Product
Porcine Prolactin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of porcine Prolactin in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA3884-48