ContactUsLinkComponent
Mouse Parathyroid Hormone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitati ve determination of mouse Parathyroid Hormone in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3491-48
Supplier:  Antibodies.com
New Product
Mouse Parathyroid Hormone ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitati ve determination of mouse Parathyroid Hormone in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3491-48
Supplier:  Antibodies.com