ContactUsLinkComponent
Rat Tyrosine Hydroxylase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Tyrosine Hydroxylase in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3896-48
Supplier:  ANTIBODIES.COM
New Product
Rat Tyrosine Hydroxylase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of rat Tyrosine Hydroxylase in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3896-48
Supplier:  ANTIBODIES.COM