Rat Fatty Acid Synthase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative de termination of rat Fatty Acid Synthase in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4746-48
Supplier: Antibodies.com