ContactUsLinkComponent
Human GPR182 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GPR182 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5420-48
New Product
Human GPR182 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human GPR182 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA5420-48