ContactUsLinkComponent
Human BST2/Tetherin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human BST2/Tetherin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5582-48
Supplier:  Antibodies.com
New Product
Human BST2/Tetherin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human BST2/Tetherin in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5582-48
Supplier:  Antibodies.com