ContactUsLinkComponent
Human SECTM1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SECTM1 in tissue homogenates, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5296-48
Supplier:  Antibodies.com
New Product
Human SECTM1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SECTM1 in tissue homogenates, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5296-48
Supplier:  Antibodies.com