ContactUsLinkComponent
Porcine MMP3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine MMP3 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3935-96
Supplier:  ANTIBODIES.COM
New Product
Porcine MMP3 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of porcine MMP3 in serum, plasma or other biological fluids. 1 * 96 Tests
Catalog # ANTIA3935-96
Supplier:  ANTIBODIES.COM