ContactUsLinkComponent
Mouse Egfl7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Egfl7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5411-48
New Product
Mouse Egfl7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Egfl7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA5411-48