ContactUsLinkComponent
Furaltadone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Furaltadone in ho ney, tissue, casing, liver. 1 * 50 Tests
Catalog # ANTIA327213-50
Supplier:  Antibodies.com
New Product
Furaltadone Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualitative determination of Furaltadone in ho ney, tissue, casing, liver. 1 * 50 Tests
Catalog # ANTIA327213-50
Supplier:  Antibodies.com