ContactUsLinkComponent
Rat KAT1/HAT1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat KAT1/HAT1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA3727-48
New Product
Rat KAT1/HAT1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinatio n of rat KAT1/HAT1 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA3727-48