ContactUsLinkComponent
Mouse Serum Amyloid A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Serum Amyloid A2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA5471-48
Supplier:  Antibodies.com
New Product
Mouse Serum Amyloid A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of mouse Serum Amyloid A2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or o 1 * 48 Tests
Catalog # ANTIA5471-48
Supplier:  Antibodies.com