ContactUsLinkComponent
Human DAOA/G72 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human DAOA/G72 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5522-48
Supplier:  Antibodies.com
New Product
Human DAOA/G72 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of human DAOA/G72 in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5522-48
Supplier:  Antibodies.com