ContactUsLinkComponent
Rat IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of rat IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4970-48
Supplier:  Antibodies.com
New Product
Rat IL-22 RA2/IL-22BP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of rat IL-22 RA2/IL-22BP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA4970-48
Supplier:  Antibodies.com