ContactUsLinkComponent
Streptomycin + Spectinomycin + Kanamycin + Neomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualit ative determination of Streptomycin + Spectinomycin + Kanamycin + Neomycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327249-96
Supplier:  ANTIBODIES.COM
New Product
Streptomycin + Spectinomycin + Kanamycin + Neomycin Strip Test Kit is a Competitive Rapid Lateral Flow Assay designed for the qualit ative determination of Streptomycin + Spectinomycin + Kanamycin + Neomycin in raw commingled cow&#0146 s milk. 1 * 96 Tests
Catalog # ANTIA327249-96
Supplier:  ANTIBODIES.COM