ContactUsLinkComponent
Human F-Spondin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human F-Spondin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5263-48
Supplier:  Antibodies.com
New Product
Human F-Spondin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human F-Spondin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5263-48
Supplier:  Antibodies.com