ContactUsLinkComponent
Rat UMOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat UMOD in serum, plasma, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5209-48
Supplier:  Antibodies.com
New Product
Rat UMOD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat UMOD in serum, plasma, urine or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5209-48
Supplier:  Antibodies.com