ContactUsLinkComponent
Extended fine tip mini Pasteur pipette, 104mm 400 sterile 1s 1 * 1 items
Catalog # ALPCLW4239SAMPLE
Supplier:  Alpha Laboratories
New Product
Extended fine tip mini Pasteur pipette, 104mm 400 sterile 1s 1 * 1 items
Catalog # ALPCLW4239SAMPLE
Supplier:  Alpha Laboratories