ContactUsLinkComponent
5 ampoules de 2 mL 6 components: Gamma-HCH (Lindane) [CAS:58-89-9] 50mg/l Triethyl phosphate [CAS:78-40-0] 20mg/l Azobenzene [CAS:103-33-3] 50mg/l Ethyl-o-silicate [CAS:78-10-4] 5mg/l Triphenylarsine [CAS:603-32-7] 25mg/l Dibenzothiophene [ 1 * 5 Ampoul
Catalog # CMPLRD0423612.5
New Product
5 ampoules de 2 mL 6 components: Gamma-HCH (Lindane) [CAS:58-89-9] 50mg/l Triethyl phosphate [CAS:78-40-0] 20mg/l Azobenzene [CAS:103-33-3] 50mg/l Ethyl-o-silicate [CAS:78-10-4] 5mg/l Triphenylarsine [CAS:603-32-7] 25mg/l Dibenzothiophene [ 1 * 5 Ampoul
Catalog # CMPLRD0423612.5