ContactUsLinkComponent
4ft Logic Class II, Type A2 Biosafety Cabinet with 8inch sash, right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand, 230V, 50/60Hz, Great Britain 1 * 1 items
Catalog # LBCP302481152
Supplier:  LABCONCO
New Product
4ft Logic Class II, Type A2 Biosafety Cabinet with 8inch sash, right-side service fixture, UV, right and left side Vacu-Pass Portals and base stand, 230V, 50/60Hz, Great Britain 1 * 1 items
Catalog # LBCP302481152
Supplier:  LABCONCO