16 components: 10mg/l each of Au B Bi Ce Ge In Mo Pd Pt Rh Ru Sb Sc Sn Te Ti in HCl 5% CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 250 mL
Catalog # CMPLRM112796L25
Supplier: CUSTOM MADE CHEMICALS LAB