Human Smooth Muscle Myosin Heavy Chain 11 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vit ro quantitative determination of human Smooth Muscle Myosin Heavy Chain 11 in serum, plasma or other biological fluid 1 * 48 Tests
Catalog # ANTIA7419-48
Supplier: Antibodies.com