ContactUsLinkComponent
Human PRCP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human PRCP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7770-48
Supplier:  Antibodies.com
New Product
Human PRCP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination o f human PRCP in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA7770-48
Supplier:  Antibodies.com