ContactUsLinkComponent
Human APMAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human APMAP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7458-48
New Product
Human APMAP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human APMAP in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA7458-48