ContactUsLinkComponent
Mouse Relaxin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of mouse Relaxin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6187-48
Supplier:  Antibodies.com
New Product
Mouse Relaxin ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determina tion of mouse Relaxin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6187-48
Supplier:  Antibodies.com