ContactUsLinkComponent
Rat Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of rat Myosin Light Chain Kinase/MLCK in tissue homogenates, cell lysates or other biological 1 * 96 Tests
  ANTIA7616-96
New Product
Rat Myosin Light Chain Kinase/MLCK ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quan titative determination of rat Myosin Light Chain Kinase/MLCK in tissue homogenates, cell lysates or other biological 1 * 96 Tests
  ANTIA7616-96