ContactUsLinkComponent
Human Calpain 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Calpain 2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6835-48
Supplier:  Antibodies.com
New Product
Human Calpain 2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human Calpain 2 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6835-48
Supplier:  Antibodies.com