ContactUsLinkComponent
Human Glutathione Peroxidase 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Glutathione Peroxidase 4 in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 48 Tests
Catalog # ANTIA8019-48
Supplier:  Antibodies.com
New Product
Human Glutathione Peroxidase 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantita tive determination of human Glutathione Peroxidase 4 in serum, plasma, tissue homogenates, cell lysates, cell culture 1 * 48 Tests
Catalog # ANTIA8019-48
Supplier:  Antibodies.com