ContactUsLinkComponent
Human UBA52 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human UBA52 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7828-96
Supplier:  ANTIBODIES.COM
New Product
Human UBA52 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human UBA52 in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA7828-96
Supplier:  ANTIBODIES.COM