ContactUsLinkComponent
Human HMBS/PBGD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human HMBS/PBGD in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6698-48
New Product
Human HMBS/PBGD ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of human HMBS/PBGD in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
  ANTIA6698-48