ContactUsLinkComponent
Human LSDP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LSDP5 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Katalogové číslo ANTIA6683-96
Dodavatel:  ANTIBODIES.COM
New Product
Human LSDP5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LSDP5 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Katalogové číslo ANTIA6683-96
Dodavatel:  ANTIBODIES.COM