ContactUsLinkComponent
Human Casein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Casein in tissue homogenates, cell lysates, cell culture supernates, breast milk or other biological fluids 1 * 48 Tests
Catalog # ANTIA8102-48
Supplier:  Antibodies.com
New Product
Human Casein ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Casein in tissue homogenates, cell lysates, cell culture supernates, breast milk or other biological fluids 1 * 48 Tests
Catalog # ANTIA8102-48
Supplier:  Antibodies.com