ContactUsLinkComponent
Human Desmoglein 3/PVA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Desmoglein 3/PVA in tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA6603-48
Supplier:  Antibodies.com
New Product
Human Desmoglein 3/PVA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative det ermination of human Desmoglein 3/PVA in tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA6603-48
Supplier:  Antibodies.com