ContactUsLinkComponent
Rat Phospholipase A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Phospholipase A2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6899-48
New Product
Rat Phospholipase A2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of rat Phospholipase A2 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA6899-48