ContactUsLinkComponent
Human Hepatitis B Virus Surface Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of human Hepatitis B Virus Surface Antigen in serum, plasma, tissue homogenates, and other 1 * 96 Tests
Catalog # ANTIA76052-96
Supplier:  ANTIBODIES.COM
New Product
Human Hepatitis B Virus Surface Antigen ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of human Hepatitis B Virus Surface Antigen in serum, plasma, tissue homogenates, and other 1 * 96 Tests
Catalog # ANTIA76052-96
Supplier:  ANTIBODIES.COM