ContactUsLinkComponent
Human AC5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AC5 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8065-96
Supplier:  ANTIBODIES.COM
New Product
Human AC5 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human AC5 in tissue homogenates, cell lysates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA8065-96
Supplier:  ANTIBODIES.COM