ContactUsLinkComponent
Mouse Glucose Transporter GLUT1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Glucose Transporter GLUT1 in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
Catalog # ANTIA8007-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Glucose Transporter GLUT1 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Glucose Transporter GLUT1 in tissue homogenates, cell lysates or other biological fluids 1 * 48 Tests
Catalog # ANTIA8007-48
Supplier:  ANTIBODIES.COM